Skip to Content
Merck
All Photos(1)

Key Documents

940208

Sigma-Aldrich

Mesocarbon microbeads

new

for lithium-ion battery anodes

Synonym(s):

Graphite, Synthetic graphite microbeads

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12171603

grade

battery grade

Quality Level

form

powder

mol wt

12.01 g/mol

composition

C

particle size

~11 μm (D50)

mp

3550 °C (lit.)
3652-3697 °C (lit.)

density

~1.2 g/cm3 (tap)

application(s)

battery manufacturing

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mesocarbon microbeads (MCMB) are a form of synthetic graphite, produced from petroleum pitch or coal tar. These materials are characterized by their distinct bead-like morphology and are categorized under "soft" carbons due to their graphitization onset at temperatures above 2000 °C. Our MCMB is synthesized to achieve a small average particle size of ~11 microns and low surface area (~1.5 m2/g).

Application

As a carbon anode material for lithium-ion batteries, MCMB exhibits several advantageous properties. It boasts a high specific capacity and its spherical morphology contributes to an increased packing density within the anode structure. The minimized surface area of MCMB plays a pivotal role in reducing detrimental side reactions throughout the charge-discharge cycles, curtailing irreversible capacity losses commonly attributed to solid-electrolyte interphase (SEI) layer formation. While MCMB has garnered recognition as an optimal anode constituent in various energy storage applications—including lithium-ion batteries, lithium-ion supercapacitors, Li-S batteries, and MCMB–Si composite batteries—it is generally deemed unsuitable for sodium-ion battery technology.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service