Skip to Content
Merck
All Photos(1)

Key Documents

941530

Poly(butylene succinate-co-butylene adipate)

new

average Mw 40 kDa

Synonym(s):

PBSA

Sign Into View Organizational & Contract Pricing


About This Item

form

powder, chunks or granules

Quality Level

mol wt

average Mw 40 kDa

color

white to yellow-brown

General description

Poly(butylene succinate-co-butylene adipate) (PBSA) is a combination of 1,4-butane diol, succinic acid, and adipic acid. It is a biodegradable, aliphatic polyester polymer that has properties similar to polypropylene and LDPE. PBSA is a sustainable replacement for petrochemical-derived plastics as it can be produced from renewable sources such as biomass. The ester group in the polymer backbone is hydrolytically degradable which makes this polymer advantageous in many applications and fields including sustainable packaging, agriculture, and medicine.

Application

  • Recyclable and Sustainable Packaging - including food packaging, films, bags, or boxes
  • Cosmetics
  • Drug delivery and implants
  • Better film forming application

Features and Benefits

  • Faster degradation
  • Thermoplastic processability
  • Sustainable alternative to petroleum based polymers
  • Excellent biodegradability

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service