Key Documents
BMSZ32615A5
Benchmark Adapter pack for 5ml tubes in Sprint 8 Plus rotors
About This Item
UNSPSC Code:
41103911
NACRES:
SB.56
manufacturer/tradename
Benchmark Scientific Z326-15-A5
General description
Adapter for 5ml tubes in Sprint 8 Plus Rotor
Application
Centrifugation of clinical samples including urine and blood. Ideal for production of PPP and PRP.
Choose from one of the most recent versions:
Certificates of Analysis (COA)