BR780421
BRAND® microcentrifuge tubes, 1.5mL with lid, polypropylene

yellow, RCF max 20k G, BIO-CERT® PCR QUALITY, 500EA
Synonym(s):
1.5mL tube, 780421, BRAND tube, centrifuge tube, microcentrifuge tube, microfuge tube, microtube, sample tube, tube, yellow tube
About This Item
material
yellow polypropylene tube
sterility
non-sterile
Sterilization Compatibility
autoclavable compatible
feature
PCR clean
RNase and DNase free
endotoxin free
pyrogen free
with cap
packaging
pack of 500 ea
manufacturer/tradename
BRAND 780421
parameter
20000 × g max. RCF
working volume
1.5 mL
Looking for similar products? Visit Product Comparison Guide
General description
The disposable microtubes can be centrifuged to the maximum RCF value stated and autoclaved at 121 °C (2 bar) according to DIN EN 285. For a reading of the approximate volume, they have an easy-to-read graduation. For easy identification of samples, different colors are available.
Features and Benefits
- BIO-CERT® PCR QUALITY: Free of DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
- Tight sealing snap lid
- Can be opened and closed with one hand
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service