Skip to Content
Merck
All Photos(1)

Key Documents

BR780424

BRAND® microcentrifuge tubes, 1.5mL with lid, polypropylene

new

orange, RCF max 20k G, BIO-CERT® PCR QUALITY, 500EA

Synonym(s):

1.5mL tube, 780424, BRAND tube, centrifuge tube, microcentrifuge tube, microfuge tube, microtube, orange tube, sample tube, tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

orange polypropylene tube

sterility

non-sterile

Sterilization Compatibility

autoclavable compatible

feature

PCR clean
RNase and DNase free
endotoxin free
pyrogen free
with cap

packaging

pack of 500 ea

manufacturer/tradename

BRAND 780424

parameter

20000 × g max. RCF

working volume

1.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

The disposable microtubes with 1.5 ml nominal volume are made from high-quality PP. The quality level BIO-CERT® PCR QUALITY ensures the required cleanliness for PCR-setups, storage of PCR samples and other applications with high cleanliness requirements.

The disposable microtubes can be centrifuged to the maximum RCF value stated and autoclaved at 121 °C (2 bar) according to DIN EN 285. For a reading of the approximate volume, they have an easy-to-read graduation. For easy identification of samples, different colors are available.

Features and Benefits

  • BIO-CERT® PCR QUALITY: Free of DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
  • Tight sealing snap lid
  • Can be opened and closed with one hand

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service