BR782713
BRAND® 2in1 cell culture inserts
24-well, PET, 1µm pore, sterile, multipack (4x12EA), BIO-CERT® quality
Synonym(s):
2-in-1, 24-well cell culture insert, 782713, BRAND insert, cell culture membrane, insert, permeable support, transwell
About This Item
Recommended Products
material
PET membrane
sterility
sterile
packaging
pack of 48 ea (4x12 multipack)
manufacturer/tradename
BRAND 782713
wells
24
pore size
1 μm pore size
binding type
cell culture treated
General description
Features and Benefits
- Can be used in hanging or standard positions
- Compatible with all common multiwell plates
- Suitable for 2D and 3D cell culture
- PET membrane with cellGrade plus surface
- Sterility Assurance Level 10-6
- BIO-CERT® cell culture quality: Free of DNA, RNase, DNase, Pyrogens, and non-cytotoxic
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)