Skip to Content
Merck
All Photos(1)

Key Documents

BR782714

BRAND® 2in1 cell culture inserts

new

24-well, PET, 3µm pore, sterlie, 48EA, BIO-CERT® quality

Synonym(s):

2-in-1, 24-well cell culture insert, 782714, BRAND insert, cell culture membrane, insert, permeable support, transwell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

PET membrane

sterility

sterile

packaging

pack of 48 ea

manufacturer/tradename

BRAND 782714

wells

24

pore size

3 μm pore size

binding type

cell culture treated

Looking for similar products? Visit Product Comparison Guide

General description

The BRAND® 2in1 BIO-CERT® cell culture insert used for skin model production, 3D epithelium cultures, chemotaxis analyses, transmigration assays, transport and polarity studies, and more. Thanks to both supporting feet and hooks, you can position the BRAND® 2in1 BIO-CERT® cell culture insert in a standing or hanging position. Due to its protein-like structure, the cellGrade plus surface provides optimal conditions for growing primary keratinocytes and other cell types. With the practical multipacks, the right number of inserts is always close at hand. They can be opened row by row so that the remaining inserts stay sterile.

Features and Benefits

  • Can be used in hanging or standard positions
  • Compatible with all common multiwell plates
  • Suitable for 2D and 3D cell culture
  • PET membrane with cellGrade plus surface
  • Sterility Assurance Level 10-6
  • BIO-CERT® cell culture quality: Free of DNA, RNase, DNase, Pyrogens, and non-cytotoxic

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service