BR782735
BRAND® 2in1 cell culture inserts
12-well, PET, 3µm pore, sterile, multipack (4x9EA), BIO-CERT® quality
Synonym(s):
12-well cell culture insert, 2-in-1, 782735, BRAND insert, cell culture membrane, insert, permeable support, transwell
About This Item
material
PET membrane
sterility
sterile
packaging
pack of 48 ea (4x9 multipack)
manufacturer/tradename
BRAND 782735
wells
12
pore size
3 μm pore size
binding type
cell culture treated
General description
Features and Benefits
- Can be used in hanging or standard positions
- Compatible with all common multiwell plates
- Suitable for 2D and 3D cell culture
- PET membrane with cellGrade plus surface
- Sterility Assurance Level 10-6
- BIO-CERT® cell culture quality: Free of DNA, RNase, DNase, Pyrogens, and non-cytotoxic
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)