GF40865913
Stainless Steel - AISI 316 alloy, FeCr18Ni10Mo3
foil, 50 x 50mm, 0.01mm thickness, not light tested (nlt)
Synonym(s):
FE240210
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11171600
NACRES:
NA.23
Recommended Products
form
foil (not light tight (nlt))
manufacturer/tradename
Goodfellow 408-659-13
size × thickness
50 mm × 50 mm × 0.01 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service