860793P
Avanti
C20 Galactosyl(ß) Ceramide (d18:1/20:0)

Avanti Research™ - A Croda Brand
Synonym(s):
D-galactosyl-ß-1,1-N-arachidoyl-D-erythro-sphingosine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C44H85NO8
Molecular Weight:
756.15
Recommended Products
Assay
≥99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (860793P-5MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
sphingolipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO[C@@H]1OC(CO)[C@H](O)C(O)C1O)[C@H](O)\C=C\CCCCCCCCCCCCC
Application
Lipids research
Packaging
5 mL Amber Glass Screw Cap Vial (860793P-5MG)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service