Skip to Content
Merck
All Photos(1)

Key Documents

890718P

Avanti

APL-718

Synonym(s):

1-oleoyl-2-(5’-(heptadecan-9-yloxy)-5’-oxopentadecanoyl)-sn-glycerol-3-ethylphosphocholine(chloride salt), Tertiary Conical EPC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H97ClNO10P
CAS Number:
Molecular Weight:
938.73
UNSPSC Code:
12352200

biological source

synthetic

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (890718P-25MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cationic lipids

shipped in

dry ice

storage temp.

-10 to -25°C

Application

Targeted Drug Delivery

Packaging

5 mL Amber Glass Screw Cap Vial (890718P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service