Skip to Content
Merck
All Photos(1)

Key Documents

5.33298

Sigma-Aldrich

TNIK Inhibitor, KY-05009

Synonym(s):

TNIK Inhibitor, KY-05009, Traf2- and Nck-interacting Kinase Inhibitor, KY-05009, KY05009, 5-(4-Methylbenzamido)-2-(phenylamino)thiazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N4O2S
CAS Number:
Molecular Weight:
352.41
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

100 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1NC(=O)c3ccc(cc3)C)C(=O)N)Nc2ccccc2

InChI key

WCEDGRTWDSHZHF-UHFFFAOYSA-N

General description

A cell-permeable, non-toxic, 2-phenylaminothiazole derivative that acts as a highly potent, ATP competitive inhibitor of Traf2- and Nck-interacting kinase (TNIK; Ki = 100 nM, IC50 = 9 nM). Also shown to inhibit the activity of MLK1 (IC50 = 18 nM). Blocks multiple effects of TGF-β1 in A549 human lung adenocarcinoma cells, including nuclear translocation of β-catenin, TNIK binding to TCF4, phosphorylation of TCF4, and phosphorylation and nuclear translocation of Smad2/3. Significantly reduces TGF-β1 induced epithelial-to-mesenchymal transition (EMT). Reduces TGF-β1-induced migration and invasion in A549 cells and inhibits gelatinase activity of MMP2 and 9. Shown to diminish TGF-β1-induced phosphorylation of focal adhesion kinase (at Tyr925), Src (at Tyr416), and paxillin (at Tyr118) and of ERK1/2 (at Thr202/Tyr204), JNK1 (Thr183/Tyr221), and JNK2 (at Thr185/Tyr223).
TNIK inhibitor, KY-05009, is a cell-permeable, highly potent inhibitor of TNIK (IC₅₀ = 9 nM). Blocks multiple effects of TGF-β1 in tumor cells.

Biochem/physiol Actions

Primary Target
TNIK

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kim, J., et al. 2014. PLOS One.9, e110180.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service