Skip to Content
Merck
All Photos(1)

Key Documents

AB9754-I

Sigma-Aldrich

Anti-Kisspeptin-10

Synonym(s):

KISS1, Kisspeptin-1, Kisspeptin-10, Metastasis-suppressor KISS-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

Quality Level

biological source

rabbit

antibody form

purified antibody

clone

polyclonal

mol wt

~16 kDa

purified by

affinity chromatography

species reactivity

human

isotype

IgG

accession no.

NP_002247

UniProt accession no.

General description

Metastasis-suppressor KISS-1 (UniProt: Q15726; also known as Kisspeptin-1) is encoded by the KISS1 (also known as PP5098) gene (Gene ID: 3814) in human. KISS1 is highly expressed in placenta and some expression is also observed in testis, pancreas, liver, small intestine. Low expression levels are also observed in the brain. Its expression is shown to increase in both early placentas and molar pregnancies and are reduced in choriocarcinoma cells. Its expression is reported to be higher levels in first trimester trophoblasts than at term of gestation. KISS-1 is a secreted metastasis suppressor that demonstrates inhibition of metastases formation in several types of cancers. It is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to generate the mature form that can also be cleaved into four chains known as Metastin (aa 68-121), Kisspeptin-14 (aa 108-121), Kisspeptin-13 (aa 109-12), and Kisspeptin-10 (112-121). Metastin acts as an endogenous ligand of GPR54, a GPCR. Activation of this receptor inhibits cell proliferation and cell migration that contributes to its tumor suppressor role. Kisspeptin-10 is reported to be a paracrine/endocrine regulator in fine-tuning trophoblast invasion generated by the trophoblast itself and increases intracellular Ca2+ levels in isolated first trimester trophoblasts. KISS1 expression is reported to be significantly reduced in human pancreatic cancer tissue and this reduced expression is associated with advanced disease. KISS1 also diminishes MMP9 expression by effecting reduced NF-kB binding to the promoter.

Specificity

This rabbit polyclonal antibody detects Kisspeptin. It targets an epitope within Kisspeptin-10 at the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids from the C-terminal region (Kisspeptin-10).

Application

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Kisspeptin-10 in human placenta tissue sections

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Quality

Evaluated by Western Blotting in HL-60 cell lysate.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service