Skip to Content
Merck
All Photos(2)

Key Documents

ABS460

Sigma-Aldrich

Anti-LEMD3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

LEM domain containing 3, Inner nuclear membrane protein Man1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LEMD3(23592)

General description

LEM domain-containing protein 3 (LEMD3), also known as Inner nuclear membrane protein Man1 (MAN1), is an inner nuclear membrane protein that interacts with BMP and activin-TGF-beta receptor-activated Smads and antagonizes TGF-beta signaling in human cells. Additionally, LEMD3 is shown to Interact with SMAD1, SMAD2, SMAD3 and SMAD5. Studies also show that LEMD3 binds to both the phosphorylated and the unphosphorylated R-SMADS. LEMD3 is expressed in heart, brain, placenta, lung, liver and skeletal muscle. Defects in LEMD3 are shown to cause disorders, such as osteopoikilosis, Buschke-Ollendorff syndrome (BOS), and melorheostosis (MEL).

Immunogen

KLH-conjugated linear peptide corresponding to human LEMD3.

Application

Anti-LEMD3 Antibody is a highly specific rabbit polyclonal antibody, that targets LEM domain-containing protein 3 & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected LEMD3 in human colon tissue.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in human colon tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected LEMD3 in 15 µg of human colon tissue lysate.

Target description

~120 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human colon tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service