Skip to Content
Merck
All Photos(1)

Key Documents

SCT052

Sigma-Aldrich

BioTracker Far-Red Peroxynitrite Live Cell Dye

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171501
NACRES:
NA.32

type

for cell culture

technique(s)

cell based assay: suitable

detection method

fluorometric

General description

Reactive oxygen species (ROS) and nitrogen species (RNS) are implicated in various physiological and pathological processes. Among ROS and RNS, peroxynitrite (ONOO−) has attracted much special attention because of the key roles that it plays in signal transduction and antimicrobial activities. Peroxynitrite is endogenously produced by reaction of nitric oxide (NO) and superoxide radical anion (O2), however, ONOO− is more cytotoxic than NO or O2 alone, contributing to numerous pathologies such as cardiovascular disease and injury, diabetes, and cancer.

The BioTracker Far-Red Peroxynitrite live cell dye can be used for ONOO- detection. This probe is cell permeable and displays a high sensitivity and selectivity towards peroxynitrite over other ROS/RNS.
This probe can be used to detect exogenous and endogenous peroxynitrite in living systems and displays long wavelength absorption (600 nm) and emission (638 nm) bands.

Spectral Properties
Absorption: 600 nm
Emissions: 638 nm

References
Wu D et al. A Far-Red-Emitting Fluorescence Probe for Sensitive and Selective Detection of Peroxynitrite in Live Cells and Tissues. Anal Chem. 2017, 89, 10924-10931.

Application

Live cell fluorescent imaging

Quality

Purity: ≥ 98% confirmed by HNMR, LC-MS and HPLC and elemental analysis
Molar Mass: 480.60 g/mol

Physical properties

480.60 g/mol

Physical form

Lyophilized

Storage and Stability

Store BioTracker Far red Peroxynitrite Live Cell Dye at -20°C, desiccate and protect from light Note: Centrifuge vial briefly to collect contents at bottom of vial before opening.

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service