Skip to Content
Merck
All Photos(1)

Key Documents

BMSNS1010E

UV-Vis spectrophotometer

new

AC/DC input 230 V, EuroPlug

Synonym(s):

Benchmark SmartDrop X Nano Spectrophotometer, Spectral Scanner, spectrometer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

manufacturer/tradename

Benchmark Scientific NS1010-E

Looking for similar products? Visit Product Comparison Guide

General description

The Accuris SmartDrop series of nano spectrophotometers provides quick and easy quantification of small samples. Utilizing a small sample volume eliminates the need for dilutions, providing more accurate results.A full color, 7” touchscreen with icon-driven menus and preprogrammed protocols gets users up and running quickly. No external computer is required. Each unit features a quartz pedestal onto which a drop of sample is placed. The arm is lowered and the drop is stretched into a liquid column of specificed length for spectral analysis. DNA, RNA and protein concentrations and purity numbers are displayed in a matter of seconds.The resulting data can be printed, saved in the SmartDrop′s onboard memory or exported via USB as an Excel file. All SmartDrop models include an integrated printer. For OD600 readings, each model includes a cuvette port and two optical glass cuvettes. Three SmartDrop models are available to meet a range of applications and budgets.A full spectrum UV-Vis spectrophotometer, SmartDrop X requires 0.5 to 2.0µl of sample. In addition to nucleic acid and protein calculations, the software has preset programs for BCA, Bradford and Lowry assays. A full spectrum analysis can be performed in about 6 seconds, aiding in theidentification of unknowns. Up to 5 specific points on the spectrum can be selected for colorimetric readings output. The long life Xenon lamp source requires no warm up and is paried with a highly sensitive CCD detector. To provide the most accurate results, the SmartDrop X utilizes 2 path

Application

Spectrophotometry, Colorimetry, OD600, UV-Vis Spectral Scanning

Features and Benefits

  • Quick and accurate results from a sample droplet
  • Touch screen control panel, no PC or software required
  • Built-in printer and cuvette port for OD600 readings


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service