Skip to Content
Merck
All Photos(2)

Key Documents

CLS3234

Corning® CellCube® T-Tubing Manifold

Medium Filter, 2 Aseptic Connectors, sterile, bag of 1 ea, case of 4 ea

Synonym(s):

lab equipment accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

material

polystyrene

sterility

sterile

feature

filter medium

packaging

bag of 1 ea
case of 4 ea

manufacturer/tradename

Corning 3234

Related Categories

Application

Adherent Cell Scale Up

Features and Benefits

  • CellCube T-Tubing
  • Med Filter
  • 2 Aseptic Connectors

Legal Information

CELLCUBE is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service