DUO87010
Duolink® PLA® Probe Mouse Anti-PhosTyr MINUS from mouse
Protein Phosphorylation Detection by Proximity Ligation Assay
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody (Protein G-Sepharose)
antibody product type
primary antibodies
clone
4G10®, monoclonal
product line
Duolink®
species reactivity
all
technique(s)
proximity ligation assay: suitable
suitability
suitable for brightfield
suitable for fluorescence
shipped in
wet ice
storage temp.
2-8°C
General description
Anti-Phosphotyrosine antibody, clone 4G10® with the Duolink® PLA® MINUS oligonucleotide. This antibody detects tyrosine phosphorylated proteins in all species. Protein target specificity can be obtained by pairing with an antibody specific to a protein of interest in a Duolink PLA experiment. This product must be used in combination with a PLUS PLA probe of a different species or another directly-conjugated primary antibody with a PLUS PLA oligo. The Anti-mouse PLA probe MINUS (DUO92001) is not necessary when using this product.
Specificity
This directly-conjugated primary antibody PLA probe recognizes tyrosine-phosphorylated proteins from all species
Application
Duolink® proximity ligation assay(PLA®) allows for endogenous detection of protein interactions, post translational modifications, and protein expression levels at the single molecule level in fixed cells and tissue samples.
This product can be applied to both the Duolink® In Situ Fluorescence Protocol and the Duolink® In Situ Brightfield Protocol depending on the detection reagents used.
Visit our Duolink® PLA Resource Center for information on how to run a Duolink® experiment, applications, troubleshooting, and more.
To perform a complete Duolink® PLA in situ experiment you will need two primary antibodies (PLA, IHC, ICC or IF validated) that recognize two target epitopes. Other necessary reagents include a pair of PLA probes from different species (one PLUS and one MINUS), detection reagents, wash buffers, and mounting medium. Note that the primary antibodies must come from the same species as the Duolink® PLA probes. Analysis is carried out using standard immunofluorescence assay equipment.HRP is also available for brightfield detection.
Application Note
Protein phosphorylation is considered as one of the key steps in signal transduction and regulation of enzymatic activity. Anti-phosTyr antibodies are commonly used in western blots after the targeted proteins have been immunoprecipitated to measure the tyrosine phosphorylation of the protein of interest. This process is time-consuming and cannot provide subcellular localization.
Anti-phosTyr PLA probes are used to detect the phosphorylation of tyrosine residues in proteins. Protein target specificity can be obtained by pairing with an antibody specific to a protein of interest in a Duolink® PLA® experiment. This product is particularly useful when site-specific anti-phosphorylation antibodies are not available. A PLUS PLA probe of a different species or another directly-conjugated primary antibody with a PLUS PLA oligo must be used in combination with this product. The Anti-mouse PLA probe MINUS (DUO92001) is not necessary when using this product. See our Product Selection Guide for more information.
Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects
This product can be applied to both the Duolink® In Situ Fluorescence Protocol and the Duolink® In Situ Brightfield Protocol depending on the detection reagents used.
Visit our Duolink® PLA Resource Center for information on how to run a Duolink® experiment, applications, troubleshooting, and more.
To perform a complete Duolink® PLA in situ experiment you will need two primary antibodies (PLA, IHC, ICC or IF validated) that recognize two target epitopes. Other necessary reagents include a pair of PLA probes from different species (one PLUS and one MINUS), detection reagents, wash buffers, and mounting medium. Note that the primary antibodies must come from the same species as the Duolink® PLA probes. Analysis is carried out using standard immunofluorescence assay equipment.HRP is also available for brightfield detection.
Application Note
Protein phosphorylation is considered as one of the key steps in signal transduction and regulation of enzymatic activity. Anti-phosTyr antibodies are commonly used in western blots after the targeted proteins have been immunoprecipitated to measure the tyrosine phosphorylation of the protein of interest. This process is time-consuming and cannot provide subcellular localization.
Anti-phosTyr PLA probes are used to detect the phosphorylation of tyrosine residues in proteins. Protein target specificity can be obtained by pairing with an antibody specific to a protein of interest in a Duolink® PLA® experiment. This product is particularly useful when site-specific anti-phosphorylation antibodies are not available. A PLUS PLA probe of a different species or another directly-conjugated primary antibody with a PLUS PLA oligo must be used in combination with this product. The Anti-mouse PLA probe MINUS (DUO92001) is not necessary when using this product. See our Product Selection Guide for more information.
Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects
Features and Benefits
- No overexpression or genetic manipulation required
- High specificity (fewer false positives)
- Single molecule sensitivity due to rolling circle amplification
- Relative quantification possible
- No special equipment needed
- Quicker and simpler than FRET
- Increased accuracy compared to co-IP
- Publication-ready results
Legal Information
4G10 is a registered trademark of Upstate Group, Inc.
Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service