Skip to Content
Merck
All Photos(2)

Key Documents

PB10100707UK

Pop-Bio ′Vü-F/C Imaging System Combo (both systems)

Synonym(s):

Imaging system, Pop-Bio ′Vü-F/C Imaging System Combo (both systems)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600

AC/DC input

24 V, UK plug

packaging

pkg of 1 ea

Looking for similar products? Visit Product Comparison Guide

Application

Western blot and fluorescence gels

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service