Skip to Content
Merck
All Photos(1)

Key Documents

SAB1410076

Sigma-Aldrich

Anti-STAM2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DKFZp564C047, Hbp, STAM2A, STAM2B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 58.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAM2(10254)

General description

The protein encoded by this gene is closely related to STAM, an adaptor protein involved in the downstream signaling of cytokine receptors, both of which contain a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). Similar to STAM, this protein acts downstream of JAK kinases, and is phosphorylated in response to cytokine stimulation. This protein and STAM thus are thought to exhibit compensatory effects on the signaling pathway downstream of JAK kinases upon cytokine stimulation. (provided by RefSeq)

Immunogen

STAM2 (AAH28740.1, 1 a.a. ~ 525 a.a) full-length human protein.

Sequence
MPLFTANPFEQDVEKATNEYNTTEDWSLIMDICDKVGSTPNGAKDCLKAIMKRVNHKVPHVALQALTLLGACVANCGKIFHLEVCSRDFATEVRAVIKNKAHPKVCEKLKSLMVEWSEEFQKDPQFSLISATIKSMKEEGITFPPAGSQTVSAAAKNGTSSNKNKEDEDIAKAIELSLQEQKQQHTETKSLYPSSEIQLNNKVARKVRALYDFEAVEDNELTFKHGEIIIVLDDSDANWWKGENHRGIGLFPSNFVTTNLNIETEAAAVDKLNVIDDDVEEIKKSEPEPVYIDEDKMDRALQVLQSIDPTDSKPDSQDLLDLEDICQQMGPMIDEKLEEIDRKHSELSELNVKVLEALELYNKLVNEAPVYSVYSKLHPPAHYPPASSGVPMQTYPVQSHGGNYMGQSIHQVTVAQSYSLGPDQIGPLRSLPPNVNSSVTAQPAQTSYLSTGQDTVSNPTYMNQNSNLQSATGTTAYTQQMGMSVDMSSYQNTTSNLPQLAGFPVTVPAHPVAQQHTNYHQQPLL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service