Skip to Content
Merck
All Photos(4)

Key Documents

SAB4702029

Monoclonal Anti-IgG (Fc) FITC produced in mouse

new

Synonym(s):

Immunoglobulin G Fc fragment

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

FITC conjugate

Quality Level

antibody product type

primary antibodies

clone

EM-07, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

0.1 mg/ml

technique(s)

flow cytometry: suitable

isotype

IgG1

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) is a 150 kDa soluble protein that serves as a major effector molecule of the humoral immune response in man. Its concentration in blood plasma of healthy individuals is approximately 10 g/l, which accounts for about 75% of the total plasma immunoglobulins. IgG has the highest stability of blood immunoglobulins (T1/2 = 21 days) and is able of placental transfer. IgG is secreted by plasma cells at a comparably high rate as other immunoglobulins.

Specificity

The mouse monoclonal antibody EM-07 reacts with Fc part of human IgG heavy chain and with isolated Fc fragments.

Immunogen

Fusion protein of human IgG Fc fragment.

Physical form

Stabilizing phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified antibody is conjugated with fluorescein isothiocyanate (FITC) under optimum conditions and unconjugated antibody and free fluorochrome are removed by size-exclusion chromatography.

Storage and Stability

Store at 2-8°C. Protect from prolonged exposure to light. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service