SAB5702273
Anti-Ret Antibody, clone 3U8T6, Rabbit Monoclonal
Synonym(s):
RET,CDHF12,CDHR16,HSCR1,MEN2A,MEN2B,MTC1,PTC,RET-ELE1,RET51
About This Item
biological source
rabbit
Quality Level
material
colorless
clone
3U8T6, monoclonal
form
liquid
species reactivity
human, rat, rat
concentration
1.00 mg/mL
technique(s)
immunofluorescence: 1:50 - 1:200
western blot: 1:500 - 1:1000
color
colorless
isotype
IgG
immunogen sequence
DISKDLEKMMVKRRDYLDLAASTPSDSLIYDDGLSEEETPLVDCNNAPLPRALPSTWIENKLYGMSDPNWPGESPVPLTRADGTNTGFPRYPNDSVYANWM
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... Ret(5979)
General description
Immunogen
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service