Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702922

Sigma-Aldrich

Anti-ZNF259 Antibody, clone 10V1A2, Rabbit Monoclonal

Synonym(s):

ZPR1, ZNF259, ZPR1 zinc finger

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

10V1A2, monoclonal

form

liquid

mol wt

51 kDa

species reactivity

mouse, mouse

concentration

0.38 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

DIRELVTKNPFTLGDSSNPGQTERLQEFSQKMDQIIEGNMKAHFIMDDPAGNSYLQNVYAPEDDPEMKVERYKRTFDQNEELGLNDMKTEGYEAGLAPQR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF259(8882)

General description

The protein encoded by this gene is found in the cytoplasm of quiescent cells but translocates to the nucleolus in proliferating cells. The encoded protein interacts with survival motor neuron protein (SMN1) to enhance pre-mRNA splicing and to induce neuronal differentiation and axonal growth. Defects in this gene or the SMN1 gene can cause spinal muscular atrophy. Two transcript variants encoding different isoforms have been found for this gene.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 360-459 of human ZNF259 (O75312).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service