Skip to Content
Merck
All Photos(1)

Key Documents

SAB5900002

Alpaca Anti-GFP single domain antibody, magnetic beads

new

recombinant Alpaca(VHH) Nano antibody magnetic bead suspension

Sign Into View Organizational & Contract Pricing


About This Item

biological source

alpaca

Quality Level

recombinant

expressed in Pichia pastoris

conjugate

magnetic beads conjugate

antibody product type

primary antibodies

clone

recombinant monoclonal

form

liquid

mol wt

15 kDa

technique(s)

ChIP: suitable
RIP: suitable
co-immunoprecipitation (co-IP): suitable
immunoprecipitation (IP): suitable
mass spectrometry (MS): suitable

color

black

matrix

(~30 -100um magnetic agarose beads)

isotype

VHH

capacity

binding capacity(10 μL slurry binds about 3ug-4μg of recombinant GFP.)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Fluorescent protein GFP exhibits bright green fluorescence when exposed to ultraviolet light and is frequently used as reporter and fusion tag. Anti-GFP single domain antibody agarose beads are agarose beads covalently coupled with VHH antibodies acquiring high specificity and affinity for GFP. These beads can efficiently capture and separate GFP and GFP tagged proteins, alongside with the associated proteins from cell extracts of mammal, plant, bacteria, yeast, insect, and other organisms.

Specificity

Selectively recognizes GFP, mEGFP, superfolder GFP and most common CFP and YFP variants, do not cross-react with mCherry, mRFP, dsRed, mTagBFP, mTagRFP or their most common derivatives.

Immunogen

GFP Tagged Protein

Features and Benefits

  • Anti Tag-Nano antibody beads are single-domain/variable region of heavy chain of camelid antibodies(sdAb/VHH).
  • They are directly coupled to beads covalently.
  • They have unique properties such as small size (~15kDa, 2-4nm) when compared to conventional antibodies.
  • No heavy and light chains in downstream interactions.
  • Acids and alkali resistance, tolerance to harsh conditions.
  • Short incubation time 5 – 30 minutes.
  • Low batch to batch variations.
  • Typically, antibodies are not detectable in Western Blot.

Packaging

plastic tube

Physical form

1XPBS, 0.03% sodium azide, 50% glycerol.

Other Notes

Before usage, please invert the vial several times (DO NOT VORTEX) to form beads suspension, and take out beads’ suspension using a pipette tip with bigger opening (i.e., cut the tip of a 1000 μl pipette tip with sterile scissors). Once opened, it is recommended to use paraffin film to seal the cap.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service