Skip to Content
Merck
All Photos(2)

Key Documents

SAB6010054

Sigma-Aldrich

Anti-phospho-PAK1/PAK2/PAK3 (S144+S141+S139) antibody, clone 3H12, Rabbit Monoclonal

new

recombinant, expressed in HEK293F cells

Synonym(s):

Serine/threonine-protein kinase PAK 3, Beta-PAK, Oligophrenin-3, p21-activated kinase 3, PAK-3, PAK3, OPHN3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

recombinant

expressed in HEK 293 F cells

Quality Level

conjugate

unconjugated

antibody product type

primary antibodies

clone

3H12, recombinant monoclonal

form

liquid

mol wt

150 kDa

purified by

affinity chromatography

species reactivity

human

concentration

0.53mg/ml

technique(s)

ELISA: 2000:10000
flow cytometry: 1:50-1:200
immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

-10 to -25°C

target post-translational modification

phosphorylation (pSer144+pSer141+pSer139)

General description

Serine/threonine protein kinase that plays a role in a variety of different signaling pathways including cytoskeleton regulation, cell migration, or cell cycle regulation. Plays a role in dendrite spine morphogenesis as well as synapse formation and plasticity. Acts as downstream effector of the small GTPases CDC42 and RAC1. Activation by the binding of active CDC42 and RAC1 results in a conformational change and a subsequent autophosphorylation on several serine and/or threonine residues. Phosphorylates MAPK4 and MAPK6 and activates the downstream target MAPKAPK5, a regulator of F-actin polymerization and cell migration. Additionally, phosphorylates TNNI3/troponin I to modulate calcium sensitivity and relaxation kinetics of thin myofilaments. May also be involved in early neuronal development.

Immunogen

A synthesized peptide derived from human Phospho-PAK1/PAK2/PAK3 (S144+S141+S139)

Sequence

YM-pS-FT

Physical form

Rabbit IgG in phosphate buffered saline , pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Storage and Stability

Upon receipt, store at -20°C or -80°C. Avoid repeated freeze.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service