Skip to Content
Merck
All Photos(1)

Key Documents

SBR00075

Sigma-Aldrich

Avibactam Sodium Salt

≥97% (HPLC), powder

Synonym(s):

NXL-104

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3NaO6S
CAS Number:
Molecular Weight:
288.23
UNSPSC Code:
51101500

Quality Level

Assay

≥97% (HPLC)

form

powder

storage condition

under inert gas (Nitrogen)

color

white

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

enzyme | inhibits

shipped in

ambient

storage temp.

−20°C

SMILES string

C1C[C@H](N2C[C@@H]1N(C2=O)OS(=O)(=O)[O-])C(=O)N.[Na+]

InChI

InChI=1S/C7H11N3O6S.Na/c8-6(11)5-2-1-4-3-9(5)7(12)10(4)16-17(13,14)15;/h4-5H,1-3H2,(H2,8,11)(H,13,14,15);/q;+1/p-1/t4-,5+;/m1./s1

InChI key

RTCIKUMODPANKX-JBUOLDKXSA-M

General description

Avibactam sodium is a β-lactamase inhibitor and β-Lactamases are responsible for resistance to penicillins, monobactams, carbapenems, and extended-spectrum cephalosporins. It is a synthetic molecule with activity against gram-negative bacteria.

Application

Avibactam and related compounds have been studied for their ability to inhibit a variety of beta-lactamases, which are enzymes that inactivate beta-lactam antibiotics such as penicillin. The development of new beta-lactamase inhibitors like Avibactam is important for combating antibiotic resistance.

Biochem/physiol Actions

Avibactam is a β-lactamase inhibitor which inhibits the activity of β-lactamases by binding to the active site of β-lactamases and blocks the access of the substrate to the enzyme. Avibactam is effective against bacteria resistant to ceftazidime like Escherichia coli.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service