Skip to Content
Merck
All Photos(1)

Key Documents

SBR00092

Sigma-Aldrich

Temocillin Disodium

≥80% (HPLC), powder

Synonym(s):

Ipconazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2Na2O7S2
CAS Number:
Molecular Weight:
458.42
UNSPSC Code:
51101500

Quality Level

Assay

≥80% (HPLC)

form

powder

color

white to yellow

antibiotic activity spectrum

Gram-negative bacteria

Mode of action

cell wall synthesis | inhibits

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC1([C@@H](N2[C@H](S1)[C@@](C2=O)(NC(=O)C(C3=CSC=C3)C(=O)O)OC)C(=O)O)C

InChI

1S/C18H24ClN3O/c1-13(2)17-8-5-15(9-14-3-6-16(19)7-4-14)18(17,23)10-22-12-20-11-21-22/h3-4,6-7,11-13,15,17,23H,5,8-10H2,1-2H3

InChI key

QTYCMDBMOLSEAM-UHFFFAOYSA-N

General description

Temocillin disodium is an aminoglycoside antibiotic which inhibits protein synthesis.

Biochem/physiol Actions

Temocillin Disodium inhibits bacterial cell wall synthesis, which is necessary for the growth and survival of bacterial cells. Specifically, it interferes with the synthesis of peptidoglycan, the major component of the bacterial cell wall, by binding to a specific target on the cell wall synthesis enzyme called penicillin-binding protein 3 (PBP3). This blocks the synthesis of peptidoglycan, leading to the weakening and eventual lysis of the bacterial cell.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service