Skip to Content
Merck
All Photos(2)

Key Documents

SML3694

Sigma-Aldrich

FR901464

≥95% (HPLC)

Synonym(s):

(2Z,4S)-4-(acetyloxy)-N-[(2R,3R,5S,6S)-6-[(2E,4E)-5-[(3R,4R,5R,7S)-4,7-dihydroxy-7-methyl-1,6-dioxaspiro[2.5]oct-5-yl]-3-methyl-2,4-pentadien-1-yl]tetrahydro-2,5-dimethyl-2H-pyran-3-yl]-2-Pentenamide, FR 901464, WB 2663B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H41NO8
CAS Number:
Molecular Weight:
507.62
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

shipped in

dry ice

storage temp.

−70°C

Biochem/physiol Actions

Inhibitor of pre-mRNA splicing; potent antitumor agent.

FR901464 is a potent anticancer agent isolated from the culture broth of the bacterium Pseudomonas sp. 2663. The compound binds to the splicing factor 3b complex and inhibits pre-mRNA splicing. FR901464 causes cell cycle arrest at the G1 and G2/M phases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service