Skip to Content
Merck
All Photos(1)

Key Documents

SML3859

Cefacetrile

≥98% (HPLC)

Synonym(s):

(6R,7R)-3-(Acetoxymethyl)-7-(cyanoacetamido)-8-oxo-5-thia-1azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-Cyanoacetamidocephalosporanic acid, Cephacetrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N3O6S
CAS Number:
Molecular Weight:
339.32
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, Light yellow to light orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Broad-spectrum first generation cephalosporin antibiotic against gram-positive and gram-negative bacteria.
Cefacetrile (cephacetrile) is a broad-spectrum first generation cephalosporin antibiotic against gram-positive and gram-negative bacteria. Cefacetrile inhibits the third and last stage of bacterial cell wall synthesis by binding to specific penicillin-binding proteins (PBPs) located inside the bacterial cell wall. Cell lysis is then mediated by bacterial cell wall autolytic enzymes such as autolysins.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service