Skip to Content
Merck
All Photos(1)

Key Documents

SML3969

Sigma-Aldrich

EMA401 sodium salt

≥98% (HPLC)

Synonym(s):

(3S)-2-(2,2-Diphenylacetyl)-1,2,3,4-tetrahydro-6-methoxy-5-(phenylmethoxy)-3-iIsoquinolinecarboxylic acid, sodium salt, (3S)-EMA400 sodium salt, (3S)-PD-126,055 sodium salt, (3S)-PD-126055 sodium salt, Olodanrigan sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H28NNaO5
CAS Number:
Molecular Weight:
529.56
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally available, selective, high-affinity type-2 angiotensin II receptor (AT2R) antagonist with in vivo pain relief efficacy.



EMA401, the (3S)-enantiomer of EMA400 (PD-126,055), is a selective, high-affinity type-2 angiotensin II receptor (AT2R) antagonist (IC50 = 39.5 nM/>39 μM against 50 pM CGP 42112A for binding human AT2R/AT1R; IC50 = 39.5 nM/408 μM against 40 nM Ang II for binding rat AT2R/AT1R). EMA401, but not the AT1R antagonist losartan, is shown to inhibit capsaicin (200 nM) responses in cultured neurons of human and rat DRG (IC50  = 10 nM). EMA401 is orally available (F = 33.2%, t1/2 = 5.7 h, Cmax = 885 ng/mL post 10 mg/kg oral dosage in rats) and exhibits in vivo pain relief efficacy among adult male SD rats with a unilateral chronic constriction injury (CCI) of the sciatic nerve (ED50 = 10 μg/kg via i.p. using the racemate EMA400).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service