Skip to Content
Merck
All Photos(1)

Key Documents

SML4032

Sigma-Aldrich

CPSF3-IN-1

≥98% (HPLC)

Synonym(s):

3-(1,3-Ddihydro-1-hydroxy-2,1-benzoxaborol-7-yl)-N-(40- acetyl-1,10-biphenyl-3-yl) propanamide, N-(4′-Acetyl[1,1′-biphenyl]-3-yl)-1,3-dihydro-1-hydroxy-2,1-benzoxaborole-7-propanamide; benzoxaborole 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22BNO4
CAS Number:
Molecular Weight:
399.25
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC(C2=CC=C(C=C2)C(C)=O)=CC=C1)CCC3=CC=CC4=C3B(OC4)O

InChI

1S/C24H22BNO4/c1-16(27)17-8-10-18(11-9-17)20-5-3-7-22(14-20)26-23(28)13-12-19-4-2-6-21-15-30-25(29)24(19)21/h2-11,14,29H,12-13,15H2,1H3,(H,26,28)

InChI key

KANKQAZGSKQFCK-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant, potent and selective inhibitor of endonuclease activity of CPSF3 in vitro that limits transcriptional termination in cells.
CPSF3-IN-1 (3-(1,3-dihydro-1-hydroxy-2,1-benzoxaborol-7-yl)-N-(40- acetyl-1,10-biphenyl-3-yl) propenamide) is a cell penetrant, potent and selective inhibitor of endonuclease activity of CPSF3 (cleavage and polyadenylation specific factor 3) in vitro that limits transcriptional termination in cells. CPSF3-IN-1 binds to CPSF3 active site and causes transcription readthrough. It potently inhibits growth of numerous cancer cell lines (IC50 = 0.28 µM HCT116 cells).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service