Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1050

Sigma-Aldrich

Anti-LHPP Antibody, clone 3B18 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:3.6.1.1, Hlhpp, Phospholysine phosphohistidine inorganic pyrophosphate phosphatase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B18, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 29.16 kDa
observed mol wt ~30 kDa

species reactivity

rat, monkey, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LHPP(64077)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3B18 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Phospholysine phosphohistidine inorganic pyrophosphate phosphatase (LHPP).

Immunogen

His-tagged full-length recombinant human Phospholysine phosphohistidine inorganic pyrophosphate phosphatase (LHPP).

Application

Anti-LHPP, clone 3B18 ZooMAb, Cat. No. ZRB1050, is a recombinant Rabbit monoclonal antibody that specifically targets LHHP and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LHPP in lysates from rat kidney, human liver, and COS-7 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected LHPP in A431 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected LHPP in human liver tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound human recombinant LHPP with a KD of 8.6 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Phospholysine phosphohistidine inorganic pyrophosphate phosphatase (UniProt: Q9H008; also known as EC:3.6.1.1, Hlhpp, LHPP) is encoded by the LHPP gene (Gene ID: 64077) in human. LHPP is an evolutionarily conserved member of the HAD-like hydrolase superfamily with phosphatase activity. It displays broad substrate specificity and can hydrolyze imidodiphosphate, 3-phosphohistidine, and 6-phospholysine. It can also hydrolyze inorganic diphosphate, but with reduced efficiency. It has two substrate binding sites that are located in amino acids 17-19 and 54-55. Its higher expression has been reported in brain, and somewhat lower expression in liver and kidney. LHHP expression is shown to be down regulated in liver and bladder cancers and knockdown of LHPP promotes the proliferation and growth of tumor cells. Conversely, sustained hepatic expression of LHPP in a murine model of hepatocellular carcinoma is reported to reduce tumor burden and prevent the loss of liver function. Silencing of LHHP can lead to enhanced phosphorylation of Akt and p65 and up-regulates the level of Bcl-2 and cyclin D1. Two isoforms of LHPP have been described that are produced by alternative splicing. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Hindupur, SK., et al. (2018). Nature. 555(7698); 678-682; Li, Y., et al. (2019). Biosci Rep. 39(7); BSR20182270.).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service