Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1976

Sigma-Aldrich

Anti-RIG-1/DDX58 Antibody, clone 1B13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ATP-dependent RNA helicase DDX58, Antiviral innate immune response receptor RIG-I, DEAD box protein 58, EC:3.6.4.13, RIG-I-like receptor 1, RLR-1, RNA sensor RIG-1, Retinoic acid-inducible gene 1 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B13, recombinant monoclonal

description

1B13 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 106.6 kDa
observed mol wt ~110 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

mouse, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... RIGI(23586)

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B13 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects RIG-1/DDX58. It targets an epitope within 24 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal half of human RIG-1/DDX58.

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RIG-1/DDX58 in HepG2 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected RIG-1/DDX58 in MCF-7 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RIG-1/DDX58 in HepG2 cells.

Affinity Binding Assay: A representative lot of this antibody bound RIG-I/DDX58 peptide with a KD of 7.8 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Antiviral innate immune response receptor RIG-I (UniProt: O95786; also known as ATP-dependent RNA helicase DDX58, EC:3.6.4.13, DEAD box protein 58, RIG-I-like receptor 1, RLR-1, RNA sensor RIG-1, Retinoic acid-inducible gene 1 protein, RIG-1) is encoded by the RIGI (also known as DDX58) gene (Gene ID: 23586) in human. RIG-1 is an innate immune receptor that senses cytoplasmic viral nucleic acids and activates a downstream signaling cascade leading to the production of type I interferons and pro-inflammatory cytokines. It detects both positive and negative strand RNA viruses. It is also reported to detect dsRNA produced from non-self dsDNA by RNA polymerase III, such as Epstein-Barr virus-encoded RNAs. It forms a ribonucleoprotein complex with viral RNAs on which it homooligomerizes to form filaments. The homooligomerization allows the recruitment of RNF135, an E3 ubiquitin-protein ligase that activates and amplifies the RIG-I-mediated antiviral signaling in an RNA length-dependent manner through ubiquitination-dependent and -independent mechanisms.  It is usually maintained in a monomeric autoinhibited state and is activated upon binding to viral RNAs, which induces a conformational shift. Upon activation, it is reported to associate with mitochondrial antiviral signaling protein (MAVS/IPS1) that activates the IKK-related kinases TBK1 and IKBKE, which in turn phosphorylate the interferon regulatory factors IRF3 and IRF7, activating transcription of antiviral immunological genes including the IFN-α  and IFN-β. RIG-1 has two CARD domains (aa 1-87 and 92-172)s and a RLR-CTR domain (aa 794-925). The RLR CTR domain controls its homooligomerization and interaction with MAVS/IPS1. In the absence of viral infection, it is maintained as a monomer in an autoinhibited state with the CARD domains masked through intramolecular interactions with the RLR-CTR domain. Upon binding to viral RNA and ubiquitination by RNF135, a conformational change releases the autoinhibition and promotes further homooligomerization. RIG-1 is reported to be phosphorylated in resting cells and is dephosphorylated in RNA virus-infected cells. Phosphorylation at threonine 770, serine 854 and serine 855 by casein kinase 2 (CK2) results in inhibition of its activity. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Shi, Y., et al. (2017). Nat. Commun. 8; 15138; Sun, Z., et al. (2011). J. Virol. 85(2); 1036-1047; Yoneyama, M., et al. (2004). Nat. Immunol. 5(7); 730-737).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service