Skip to Content
Merck
All Photos(1)

Key Documents

BOTB9046

Supelco

Taraxacum officinale Root

Botanical reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

manufacturer/tradename

BOTANICERT®

application(s)

food and beverages
pharmaceutical

format

matrix material

storage temp.

2-25°C

General description

English name: Dandelion, French name: Pissenlit

This botanical reference material (BRM) ismanufactured by BotaniCERT™ and EXTRASYNTHESE™. The material is sourced frombotanical gardens, collected by botanical experts during the flowering period,analyzed by seasoned phytochemists and geneticians.

BotaniCERT Botanical Reference Materials are samples ofplants or plant parts (leaf, root, rhizome, fruit, flower, seed...) authenticatedby organoleptic criteria, macroscopic and microscopic examination, HP-TLC andHPLC phytochemical fingerprint and genetic markers by PCR. They are given with2 or 3 markers (ITS, matK, rbcL) and DNA sequences are reported.

The products are provided as 5g samples and a detailedCertificate of Analysis, including related pictures, chromatograms and DNAsequences is available.

Using a Botanical Reference Material in an identificationanalysis allows calibration of methods and increases the guarantee of theresult obtained.

Application

For identity and authenticity testing of botanical materials

Legal Information

BOTANICERT is a registered trademark of BOTANICERT, Société, FR

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service