Skip to Content
Merck
Monosodium Glutamate ≥99%, FG

Monosodium Glutamate

Monosodium Glutamate ≥99%, FG
Synonym(s):
L-Glutamic acid monosodium salt
Linear Formula:
NaOOCCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
169.11

Products

Brand
Feature
Formula Weight
Purity
Agency
Quality Segment
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing