Skip to Content
Merck
All Photos(1)

Key Documents

W275601

Sigma-Aldrich

Monosodium Glutamate

new

≥99%, FG

Synonym(s):

L-Glutamic acid monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
169.11
UNSPSC Code:
12352200
E Number:
E 621
grade:
FG
Halal
Kosher
food allergen:
no known allergens

grade

FG
Halal
Kosher

Quality Level

reg. compliance

EU Regulation 1333/2008 & 178/2002
FDA 21 CFR 117

Assay

≥99%

food allergen

no known allergens

Organoleptic

odorless

SMILES string

[Na+].N[C@@H](CCC(=O)O)C(=O)[O-]

InChI

1S/C5H9NO4.Na/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);/q;+1/p-1/t3-;/m0./s1

InChI key

LPUQAYUQRXPFSQ-DFWYDOINSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service