Skip to Content
Merck
Vibrio furnissii NCTC 11218 Lenticule® discs >100 CFU, reference material, suitable for microbiology

Vibrio furnissii NCTC 11218 Lenticule® discs

Vibrio furnissii NCTC 11218 Lenticule® discs >100 CFU, reference material, suitable for microbiology

Products

Brand
Physical Form
Quality Segment
Application
Microorganism Suitability