RM11218Q
Vibrio furnissii NCTC 11218 Lenticule® discs
>100 CFU, reference material, suitable for microbiology
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
85151701
NACRES:
NA.24
Recommended Products
grade
reference material
Quality Level
form
disc (hydrosoluble)
shelf life
limited shelf life, expiry date on the label
application(s)
environmental
food and beverages
microbiology
storage temp.
−20°C
suitability
Vibrio spp.
Looking for similar products? Visit Product Comparison Guide
General description
This reference material (RM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this RM can be found on the certificate of analysis.
Download your certificate of analysis at: http://www.sigma-aldrich.com.
LENTICULE® discs are manufactured by Sigma-Aldrich under licence from UK Health Security Agency.
Download your certificate of analysis at: http://www.sigma-aldrich.com.
LENTICULE® discs are manufactured by Sigma-Aldrich under licence from UK Health Security Agency.
Other Notes
Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)
Legal Information
Lenticule is a registered trademark of The Secretary of State for Health (United Kingdom)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service