주요 문서
BR4630340
BRAND® Dispensette® S Organic, Digital bottle-top dispenser
volume range 1-10 mL, without recirculation valve
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121500
NACRES:
NB.01
특징
incremental 0.05 mL
DE-M marking
without recirculation valve
제조업체/상표
BRAND 4630340
용적 범위
1-10 mL
피팅
(GL 28/S 28)
GL 32
GL 38
S 40
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Dispensette S Organic is ideal for dispensing of organic solvents and acids
특징 및 장점
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
기타 정보
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
법적 정보
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)