주요 문서
DWK227497-17G
Wheaton® Bod Bottle
capacity 300 mL, Un-numbered, bottle diam. × H 69 mm × 165 mm, pennyhead glass cap (stopper)
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.43
material
pennyhead glass cap (stopper)
포장
pkg of 24 ea
제조업체/상표
WHEATON® 227497-17G
보틀 직경 × 높이
69 mm × 165 mm
용량
300 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
BOD Bottles are ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action. We recommend the 300mL capacity bottles for the five-day BOD test as referenced in Standard Methods for the Examination of Water and Wastewater, 21st Edition, 5210B, 2005.
특징 및 장점
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
법적 정보
WHEATON is a registered trademark of Wheaton Industries, Inc.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)