주요 문서
Z303410
Plastic hose connectors for Quickfit® water-cooled condenser
동의어(들):
4510/02
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103025
NACRES:
NB.35
포장
pkg of 10 ea
제조업체/상표
Quickfit
재고 정보
available only in EU
일반 설명
These screwthread hose connectors are only for use with the QUICKFIT® and PYREX® glassware products. They do not fit DURAN® glass threads.
법적 정보
DURAN is a registered trademark of DWK Life Sciences
PYREX is a registered trademark of Corning, Inc.
QUICKFIT is a registered trademark of DWK Life Sciences
Quickfit is a registered trademark of SciLabware Limited
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)