주요 문서
Z660639
Catalyst heater for Plas-Labs temperature and humidity box
AC/DC input 240 V AC, UK plug
동의어(들):
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103406
NACRES:
NB.35
제조업체/상표
Plas-Labs 800-HEATER
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Plas-Labs catalyst heater has two primary functions for use in the Plas-Labs temperature and humidity glove box. The catalyst heater reduces trace amounts of oxygen and maintains incubation temperature levels of 35-37 °C. The internal atmosphere is drawn off the floor and up across heating elements and aluminized palladium pellets. Any oxygen present will react with your anaerobic gas mixture and be reduced to water vapor. When used in conjunction with the Plas-Labs temperature and humidity glove boxes with drying train attached, the vapor is then removed from the box as needed. CE compliant.
법적 정보
관련 제품
제품 번호
설명
가격
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)