주요 문서
Z689890
Sigma-Aldrich
Aldrich® flexible support system
with base plate, L 457 mm
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41100000
NACRES:
NB.35
길이
457 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The unique flexible arms can be placed in virtually any position or angle. Available in three different systems: with steel base plate, with alloy lab frame connector for rods up 19 mm diameter, or aluminum bench clamp.
Each system includes: three nickel-plated clamp heads, 2-prong clamp with 75 mm max. grip, 3-prong clamp with 69 mm max. grip and spring clamp with 13 mm max. grip., and one nickel-plated flex arm in choice of 305 mm length or 457 mm length.
Clamp heads with prongs include vinyl and fiberglass covers for high temperature applications. Utility wrench included with each system.
법적 정보
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)