주요 문서
Z744084
Sigma-Aldrich
Water Purifier Cartridge for H-Genie® Lite
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
12161503
NACRES:
NA.22
일반 설명
Water Purifier Cartridge for H-Genie® Lite is designed to keep the water purity at the required level to maintain optimum cell lifetime. Expected consumption with normal daily use is 6 cartridges per a year.
H-Genie® Lite water purifier cartridges are to be used with H-Genie® Lite benchtop hydrogen generator (Z744083)
법적 정보
H-Genie is a registered trademark of ThalesNano Energy
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)