모든 사진(1)
About This Item
실험식(Hill 표기법):
C12H22MgO14 · 2H2O
CAS Number:
Molecular Weight:
450.63
UNSPSC 코드:
12352200
NACRES:
NA.24
추천 제품
일반 설명
This Standard Reference Material (SRM) is certified for use as an assay standard for magnesium. It is intended primarily for use in the calibration and standardization of procedures employed in magnesium analysis and for the routine critical evaluation of daily working standards used in these procedures. The material is magnesium gluconate dihydrate: Mg (C6H11O7)2 • 2H2O and is supplied in a unit of 5 g. For more information, please refer to the COA and SDS.
SRM 929A_cert SRM 929A _SDS
SRM 929A_cert SRM 929A _SDS
법적 정보
NIST is a registered trademark of National Institute of Standards and Technology
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
가장 최신 버전 중 하나를 선택하세요:
자사의 과학자팀은 생명 과학, 재료 과학, 화학 합성, 크로마토그래피, 분석 및 기타 많은 영역을 포함한 모든 과학 분야에 경험이 있습니다..
고객지원팀으로 연락바랍니다.