BMSIPD9600BD28
Ball Dropper to Fill 96 BeadBlaster™ 96 Well Plate
diam. 2.8 mm
로그인조직 및 계약 가격 보기
크기 선택
25 MG
$87.60
$87.60
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
25 MG
$87.60
About This Item
UNSPSC 코드:
41100000
NACRES:
NA.82
$87.60
구입 가능 여부는 고객센터에 문의하십시오.
추천 제품
material
acrylic
포장
pack of 1 ea
제조업체/상표
Benchmark Scientific IPD9600-BD28
직경
2.8 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
관련 카테고리
1 of 4
이 품목 | CBR00103 | CDS005773 | CDS017300 |
---|---|---|---|
form solid | form solid | form solid | form solid |
일반 설명
This ball dropper is used to easily fill 96 well plates with 1 bead per well.
애플리케이션
Homogenization/milling/grinding of samples
법적 정보
BeadBlaster is a trademark of Benchmark Scientific, Inc.
가장 최신 버전 중 하나를 선택하세요:
Lingling Chen et al.
Scientific reports, 7(1), 12218-12218 (2017-09-25)
Optical clearing methods can facilitate deep optical imaging in biological tissue by reducing light scattering and this has enabled accurate three-dimensional signal visualization and quantification of complex biological structures. Unfortunately, existing optical clearing approaches present a compromise between maximizing clearing
R Hommel et al.
Journal of basic microbiology, 30(4), 297-300 (1990-01-01)
An improved solubilization procedure for the membrane-bound quinoprotein aldehyde dehydrogenase from Acetobacter rancens CCM 1774 was established. After the first solubilization of membrane enzymes by Brij 35 which provided important extraction of membrane proteins other than aldehyde dehydrogenase, the application
J K Towns et al.
Analytical chemistry, 63(11), 1126-1132 (1991-06-01)
Capillary zone electrophoretic separations of proteins have been achieved by using nonionic surfactant coated capillaries. Capillaries were prepared by derivatization of the silica surface with octadecylsilane followed by the deposition of a layer of nonionic surfactant from an aqueous solution
B T Dumas et al.
Clinica chimica acta; international journal of clinical chemistry, 258(1), 21-30 (1997-02-03)
A rapid and reliable method for measuring serum albumin employing bromcresol green is described. The addition of albumin to a solution of bromcresol green in a 0.075 M succinate buffer pH 4.20 results in an increase in absorbance at 628
Hitesh S Deshmukh et al.
American journal of respiratory cell and molecular biology, 38(4), 446-454 (2007-11-17)
Chronic obstructive pulmonary disease (COPD), a global public health problem, is characterized by progressive difficulty in breathing, with increased mucin production, especially in the small airways. Acrolein, a constituent of cigarette smoke and an endogenous mediator of oxidative stress, increases