주요 문서
HW11089-02
Sigma-Aldrich
Lifting Bar, 8 Loop, ∅20" Body, Angled
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41116107
NACRES:
RA.01
일반 설명
SAFC has qualified EZ BioPac, from ILC Dover as their preferred bulk powder package. Manufactured with Armorflex 114 film, the EzBiopac range is a fit for purpose, scalable and intuitive offer that enables the clean, safe and efficient delivery of bulk powders into bioprocess.
This is an accessory which can be used to assist with the handling of the EZ BioPac by ILC Dover filled with cell culture media.
기타 정보
Low profile lifting bar for products DP4487, DP4600, DP4537, DP4601, DP4652
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)