주요 문서
OH30061978GB
Ohaus® Explorer® Precision Balance
Model EX324/AD, weighing capacity 320 g, resolution: 0.1 mg, AC/DC input 220/240 V, UK plug
동의어(들):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41111502
NACRES:
NB.71
특징
CE compliant
포장
pack of 1 ea
제조업체/상표
Ohaus® 30061978GB
계량 용량 측정 범위
320 g, resolution: 0.1 mg
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The OHAUS Explorer Series of precision balances combine modern features and design elements to offer unmatched functionality in a line of high-performance balances unlike any other on the market. These smart and intuitive balances simplify even the most complex laboratory measurements.
애플리케이션
Weighing, Parts Counting, Percent Weighing, Checkweighing, Animal/Dynamic Weighing, Filling, Totalization/Statistics, Formulation, Differential Weighing, Density Determination, Peak Hold, Ingredient Costing, Pipette Adjustment, SQC, Statistics
특징 및 장점
Selectable environmental filters, auto tare, user selectable span calibration points, software lockout and reset menu, user selectable communication settings and data print options, user definable project and user IDs, auto standby, up to 14 operating languages
법적 정보
Explorer is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)