주요 문서
Z740395
The SpecTainer II
size 60 mL, non-sterile
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.22
material
polyethylene cap
polypropylene container
무균
non-sterile
포장
pack of 500 ea
제조업체/상표
Simport C567-60Y
크기
60 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
For applications needing the utmost security where sample integrity is of high importance. A great feature of the Spectainer I is that it incorporates a unique tamper evident screw cap ensuring peace of mind during transport or storage situations where someone might have tampered with the specimen.
Both containers and caps are manufactured without the use of plasticisers or mold release agents. All material used is free from latex. Containers are 95kpa compliant. Available in three sizes: 60, 90 and 120 ml. The 60 ml size however is not graduated. The sterile container protects its sterility with a tamper evident seal.
법적 정보
SpecTainer is a trademark of Simport
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)