주요 문서
25069
Supelco
TDS3 Sampling Caps with Washers for 1/4 in. Containers
greener alternative
for use with TDS3 to convert it to sampling
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41104008
포장
pkg of 1 ea
Quality Level
환경친화적 대안 제품 특성
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
기술
active air sampling: suitable
응용 분야
air monitoring
environmental
industrial hygiene
호환성
for use with TDS3 to convert it to sampling
환경친화적 대안 카테고리
일반 설명
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.
법적 정보
TDS3 is a trademark of Sigma-Aldrich Co. LLC
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)